Anti-USP31 Mouse Monoclonal Antibody [clone: 3B6]
USBI135169
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 31
- Antigen symbol:USP31
- Clonality:Monoclonal
- Clone:3B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1254 to 1353
- Storage buffer:PBS, pH 7,2.
- Sequence:AGGSSVKSVCKNTGDDEAERGHQPPASQQPNANTTGKEQLVTKDPASAKHSLLSARKSKSSQLDSGVPSSPGGRQSAEKSSKKLSSSMQTSARPSQKPQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: USP31 (ubiquitin specific peptidase 31)
Clonality: monoclonal
Clone: 3B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human