Anti-USP29 Mouse Monoclonal Antibody [clone: 1A8]
USBI135166
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 29
- Antigen symbol:USP29
- Clonality:Monoclonal
- Clone:1A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:131 to 241
- Storage buffer:PBS, pH 7,2.
- Sequence:KTSFYSICNKPSYQKMPLFMSKSPTHVKKGILENQGGKGQNTLSSDVQTNEDILKEDNPVPNKKYKTDSLKYIQSNRKNPSSLEDLEKDRDLKLGPSFNTNCNGNPNLDE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 1. 2?g/ml
Type: Primary
Antigen: USP29 (ubiquitin specific peptidase 29)
Clonality: monoclonal
Clone: 1A8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human