Anti-SSX3 Mouse Monoclonal Antibody [clone: 4A11]
Catalog # USBI133886
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Synovial sarcoma, X breakpoint 3
- Antigen symbol:SSX3
- Clonality:Monoclonal
- Clone:4A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 100
- Storage buffer:PBS, pH 7,2.
- Sequence:MNGDDTFARRPTVGAQIPEKIQKAFDDIAKYFSKEEWEKMKVSEKIVYVYMKRKYEAMTKLGFKAILPSFMRNKRVTDFQGNDFDNDPNRGNQVQRPQMT
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SSX3 (synovial sarcoma, X breakpoint 3)
Clonality: monoclonal
Clone: 4A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human