Anti-SPRR2F Mouse Monoclonal Antibody [clone: 5A9]
Catalog # USBI133802
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:small proline-rich Protein 2F
- Antigen symbol:SPRR2F
- Clonality:Monoclonal
- Clone:5A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 72
- Storage buffer:PBS, pH 7,2.
- Sequence:MSYQQQQCKQPCQPPPVCPAPKCPEPCPPPKCPEPCPPSKCPQSCPPQQCQQKCPPVTPSPPCQPKCPPKSK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SPRR2F (small proline-rich protein 2F)
Clonality: monoclonal
Clone: 5A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human