Anti-PLEK Rabbit Polyclonal Antibody
Catalog # USBI131447
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Pleckstrin
- Antigen symbol:PLEK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 350
- Storage buffer:PBS, pH 7,2.
- Sequence:MEPKRIREGYLVKKGSVFNTWKPMWVVLLEDGIEFYKKKSDNSPKGMIPLKGSTLTSPCQDFGKRMFVFKITTTKQQDHFFQAAFLEERDAWVRDINKAIKCIEGGQKFARKSTRRSIRLPETIDLGALYLSMKDTEKGIKELNLEKDKKIFNHCFTGNCVIDWLVSNQSVRNRQEGLMIASSLLNEGYLQPAGDMSKSAVDGTAENPFLDNPDAFYYFPDSGFFCEENSSDDDVILKEEFRGVIIKQGCLLKQGHRRKNWKVRKFILREDPAYLHYYDPAGAEDPLGAIHLRGCVVTSVESNSNGRKSEEENLFEIITADEVHYFLQAATPKERTEWIKAIQMASRTGK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PLEK (pleckstrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human