Anti-NKX3-1 Mouse Monoclonal Antibody [clone: 1C7]
Catalog # USBI130400
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NK3 homeobox 1
- Antigen symbol:NKX3-1
- Clonality:Monoclonal
- Clone:1C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:100 to 210
- Storage buffer:PBS, pH 7,2.
- Sequence:GSYLLDSENTSGALPRLPQTPKQPQKRSRAAFSHTQVIELERKFSHQKYLSAPERAHLAKNLKLTETQVKIWFQNRRYKTKRKQLSSELGDLEKHSSLPALKEEAFSRAS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: NKX3-1 (NK3 homeobox 1)
Clonality: monoclonal
Clone: 1C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat