Anti-NID1 Mouse Polyclonal Antibody
50 µg
Catalog # USBI130368
Supplier: US Biological
Jump to:
Product Details & Documents
Type: Primary
Antigen: NID1 (nidogen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Mouse
- Shipping temperature:Ice
- Antigen name:Nidogen 1
- Cat. No.:USBI130368
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:NID1
- Storage buffer:PBS, pH 7,2.



