Anti-NBN Mouse Monoclonal Antibody [clone: 3E4]
USBI130364
- Antibody type:Primary
- Antigen name:nibrin
- Antigen symbol:NBN
- Clonality:Monoclonal
- Clone:3E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:645 to 754
- Storage buffer:PBS, pH 7,2.
- Sequence:DDSEMLPKKLLLTEFRSLVIKNSTSRNPSGINDDYGQLKNFKKFKKVTYPGAGKLPHIIGGSDLIAHHARKNTELEEWLRQEMEVQNQHAKEESLADDLFRYNPYLKRRR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: NBN (nibrin)
Clonality: monoclonal
Clone: 30000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human