Anti-MKRN1 Mouse Monoclonal Antibody [clone: 4E9]
Catalog # USBI129707
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:makorin ring finger Protein 1
- Antigen symbol:MKRN1
- Clonality:Monoclonal
- Clone:4E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:365 to 441
- Storage buffer:PBS, pH 7,2.
- Sequence:MSNKACRYFDEGRGSCPFGGNCFYKHAYPDGRREEPQRQKVGTSSRYRAQRRNHFWELIEERENSNPFDNDEEEVV*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MKRN1 (makorin ring finger protein 1)
Clonality: monoclonal
Clone: 4000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human