Anti-IL11 Mouse Monoclonal Antibody [clone: 1F1]
USBI128383
- Antibody type:Primary
- Antigen name:Interleukin 11
- Antigen symbol:IL11
- Clonality:Monoclonal
- Clone:1F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:25 to 74
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: IL11 (interleukin 11)
Clonality: monoclonal
Clone: 1F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human