Anti-FKBP14 Mouse Polyclonal Antibody
Catalog # USBI126828
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:FK506 binding protein 14, 22 kDa
- Antigen symbol:FKBP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 211
- Storage buffer:PBS, pH 7,2.
- Sequence:MRLFLWNAVLTLFVTSLIGALIPEPEVKIEVLQKPFICHRKTKGGDLMLVHYEGYLEKDGSLFHSTHKHNNGQPIWFTLGILEALKGWDQGLKGMCVGEKRKLIIPPALGYGKEGKGKIPPESTLIFNIDLLEIRNGPRSHESFQEMDLNDDWKLSKDEVKAYLKKEFEKHGAVVNESHHDALVEDIFDKEDEDKDGFISAREFTYKHDEL
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FKBP14 (FK506 binding protein 14, 22 kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human