Anti-CST3 Rabbit Polyclonal Antibody
100 µg
Catalog # USBI125608
Supplier: US Biological
Jump to:
Product Details & Documents
Type: Primary
Antigen: CST3 (cystatin C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Amino acid number:1 to 146
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Sequence:MAGPLRAPLLLLAILAVALAVSPAAGSSPGKPPRLVGGPMDASVEEEGVRRALDFAVGEYNKASNDMYHSRALQVVRARKQIVAGVNYFLDVELGRTTCTKTQPNLDNCPFHDQPHLKRKAFCSFQIYAVPWQGTMTLSKSTCQDA
- Shipping temperature:Ice
- Antigen name:Cystatin C
- Cat. No.:USBI125608
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:CST3
- Storage buffer:PBS, pH 7,2.



