Anti-CYP4V2 Rabbit Polyclonal Antibody
USBI125597
- Antibody type:Primary
- Antigen name:cytochrome P450, family 4, subfamily V, polypeptide 2
- Antigen symbol:CYP4V2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 525
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CYP4V2 (cytochrome P450, family 4, subfamily V, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human