Anti-CFDP1 Mouse Monoclonal Antibody [clone: 5B7]
USBI124894
- Antibody type:Primary
- Antigen name:Craniofacial Development Protein 1
- Antigen symbol:CFDP1
- Clonality:Monoclonal
- Clone:5B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:168 to 251
- Storage buffer:PBS, pH 7,2.
- Sequence:VFDFAGEEVRVTKEVDATSKEAKSFFKQNEKEKPQANVPSALPSLPAGSGLKRSSGMSSLLGKIGAKKQKMSTLEKSKLDWESF
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CFDP1 (craniofacial development protein 1)
Clonality: monoclonal
Clone: 5B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human