Anti-CDH19 Mouse Monoclonal Antibody [clone: 1G4]
Catalog # USBI124771
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cadherin 19, type 2
- Antigen symbol:CDH19
- Clonality:Monoclonal
- Clone:1G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:231 to 341
- Storage buffer:PBS, pH 7,2.
- Sequence:GQPGALSGTTSVLIKLSDVNDNKPIFKESLYRLTVSESAPTGTSIGTIMAYDNDIGENAEMDYSIEEDDSQTFDIITNHETQEGIVILKKKVDFEHQNHYGIRAKVKNHH*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CDH19 (cadherin 19, type 2)
Clonality: monoclonal
Clone: 1G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human