Anti-ALOX15 Rabbit Polyclonal Antibody
Catalog # USBI123247
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Arachidonate 15-lipoxygenase
- Antigen symbol:ALOX15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:15LOX-1
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ALOX15 (arachidonate 15-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse