Anti-TNFRSF10B Mouse Monoclonal Antibody ATTO 488 [clone: HS201]
Catalog # USBI045354
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 10b
- Antigen symbol:TNFRSF10B
- Clonality:Monoclonal
- Clone:HS201
- Conjugation:ATTO 488
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: Monoclonal
Clone: HS201
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: