Anti-TNFRSF13B Rat Monoclonal Antibody (Biotin) [clone: 1A-10]
USBI045322
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 13B
- Antigen symbol:TNFRSF13B
- Clonality:Monoclonal
- Clone:1A-10
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TNFRSF13B (tumor necrosis factor receptor superfamily, member 13B)
Clonality: Monoclonal
Clone: 1A-10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: