Anti-IL-33, blocking Mouse Monoclonal Antibody [clone: Bondy-1-1]
Catalog # USBI044927
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:IL-33, blocking
- Antigen symbol:IL-33
- Clonality:Monoclonal
- Clone:Bondy-1-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Form:liquid
- Storage buffer:PBS, 10% glycerol, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IL-33, blocking
Clonality: Monoclonal
Clone: Bondy-1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: