Anti-IL17A Mouse Monoclonal Antibody [clone: 17-142F]
Catalog # USBI044903
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17A
- Antigen symbol:IL17A
- Clonality:Monoclonal
- Clone:17-142F
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Form:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: Monoclonal
Clone: 17-142F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: