Anti-GITRL Rat Monoclonal Antibody [clone: MGTL 15]
Catalog # USBI044850
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:GITRL
- Clonality:Monoclonal
- Clone:MGTL 15
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Western blot:Yes
- Form:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GITRL
Clonality: Monoclonal
Clone: MGTL 15
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: