Anti-FGF-21 Rabbit Polyclonal Antibody
100 µg
Catalog # USBI044800
Supplier: US Biological
Jump to:
Product Details & Documents
Type: Primary
Antigen: FGF-21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Documents
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Storage temperature:Cool
- Host:Rabbit
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Cat. No.:USBI044800
- Conjugation:Unconjugated
- Antigen symbol:FGF-21
- Storage buffer:0,2um-filtered PBS, pH 7,4
- ELISA:Yes



