Order Entry
Export
ContactUsLinkComponent
Anti-FGF-19 Mouse Monoclonal Antibody [clone: FG369-1]
Anti-FGF-19 Mouse Monoclonal Antibody [clone: FG369-1]
  USBI044793
Anti-FGF-19 Mouse Monoclonal Antibody [clone: FG369-1]
  USBI044793

 

  • Antibody type:
    Primary
  • Antigen symbol:
    FGF-19
  • Clonality:
    Monoclonal
  • Clone:
    FG369-1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,2um-filtered PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: FGF-19
Clonality: Monoclonal
Clone: FG369-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: