Order Entry
Export
ContactUsLinkComponent
Anti-ANGPTL7 Rabbit Polyclonal Antibody
Anti-ANGPTL7 Rabbit Polyclonal Antibody
Catalog # USBI044553
Supplier:  US Biological
Anti-ANGPTL7 Rabbit Polyclonal Antibody
Catalog # USBI044553
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Angiopoietin-like 7
  • Antigen symbol:
    ANGPTL7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,2um-filtered PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ANGPTL7 (angiopoietin-like 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: