Anti-ADIPOR1 Rabbit Polyclonal Antibody
USBI044474
- Antibody type:Primary
- Antigen name:adiponectin receptor 1
- Antigen symbol:ADIPOR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Amino acid number:2 to 36
- Storage buffer:0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: ADIPOR1 (adiponectin receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: