Anti-ADIPOQ Rabbit Polyclonal Antibody
Catalog # USBI044455
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Adiponectin
- Antigen symbol:ADIPOQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Western blot:Yes
- Form:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: