Anti-LGR5 Rabbit Polyclonal Antibody
Catalog # USBI037761
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:leucine-rich repeat containing G Protein-coupled receptor 5
- Antigen symbol:LGR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:247 to 277
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50
Type: Primary
Antigen: LGR5 (leucine-rich repeat containing G protein-coupled receptor 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human