Anti-IL31 Rabbit Polyclonal Antibody
Catalog # USBI037009
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:interleukin 31
- Antigen symbol:IL31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: IL31 (interleukin 31)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse