Anti-GCNT2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI035968-AP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
- Antigen symbol:GCNT2
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Amino acid number:212 to 240
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: GCNT2 (glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human