Anti-FGFR1 Rabbit Polyclonal Antibody
200 µl
Catalog # USBI035639
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:50-1:100; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: FGFR1 (fibroblast growth factor receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Purification:purified by Protein G chromatography
- Antibody type:Primary
- Epitope:N-terminal
- Form:liquid
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Fibroblast growth factor receptor 1
- Cat. No.:USBI035639
- Reactivity:Human|Mouse
- Conjugation:Unconjugated
- Antigen symbol:FGFR1
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- ELISA:Yes



