Anti-E2F7 Rabbit Polyclonal Antibody
200 µl
Catalog # USBI034890
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: E2F7 (E2F transcription factor 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Epitope:internal
- Form:liquid
- Amino acid number:590 to 619
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:E2F transcription factor 7
- Cat. No.:USBI034890
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:E2F7
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- ELISA:Yes



