Anti-GPATCH11 Rabbit Polyclonal Antibody
200 µl
Catalog # USBI033343
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: GPATCH11 (G patch domain containing 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Epitope:N-terminal
- Form:liquid
- Amino acid number:15 to 45
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:G Patch Domain Containing 11
- Cat. No.:USBI033343
- Reactivity:Hamster|Human|Mouse
- Conjugation:Unconjugated
- Antigen symbol:GPATCH11
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- ELISA:Yes



