Order Entry
Export
ContactUsLinkComponent
Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 13B811]
Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 13B811]
Catalog # USBI031347
Supplier:  US Biological
Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 13B811]
Catalog # USBI031347
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 eta
  • Clonality:
    Monoclonal
  • Clone:
    13B811
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:500-1:1000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: 14-3-3 eta (14-3-3 eta)
Clonality: Monoclonal
Clone: 13B811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat