Order Entry
Export
ContactUsLinkComponent
 
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 13B794]
Catalog # USBI030716
Supplier:  US Biological
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 13B794]
Catalog # USBI030716
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vascular cell adhesion molecule 1
  • Antigen symbol:
    VCAM1
  • Clonality:
    Monoclonal
  • Clone:
    13B794
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Storage buffer:
    0,03% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 13B794
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human