Anti-ACTA2 Mouse Monoclonal Antibody [clone: 13B14]
Catalog # USBI029936
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Actin, alpha 2, smooth muscle, aorta
- Antigen symbol:ACTA2
- Clonality:Monoclonal
- Clone:13B14
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: Monoclonal
Clone: 13B14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat