Anti-FST Rabbit Polyclonal Antibody
Catalog # USBI029780
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Follistatin
- Antigen symbol:FST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,5% BSA, 0,01% thimerosal, 30% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: FST (follistatin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human