Order Entry
Export
ContactUsLinkComponent
Anti-Myosin IIA, Non-muscle Rabbit Polyclonal Antibody
Anti-Myosin IIA, Non-muscle Rabbit Polyclonal Antibody
Catalog # USBI029547
Supplier:  US Biological
undefined
Anti-Myosin IIA, Non-muscle Rabbit Polyclonal Antibody
Catalog # USBI029547
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myosin 2A
  • Antigen symbol:
    MYO2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Ser1943
  • Storage buffer:
    0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:300; Immunochemistry: 1:500; Immunofluorescence: 1:400

Type: Primary
Antigen: Myosin IIA, Non-muscle
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human