Anti-SELL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: DREG-56]
Catalog # ROCK200-326-N72
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:DREG-56
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:6402
- Antigen synonyms:LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,2% BSA (w/v)
- Storage temperature:Cool
- Concentration:Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD62L Antibody (Monoclonal) was produced by repeated immunizations with CD62L antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Anti-CD62L is useful for Flow Cytometry and Western Blot. Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions:
Do not freeze!
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: DREG-56
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human