Anti-lambda light chain Goat Polyclonal Antibody
1 mg
Catalog # ROCK609-1111
Supplier: Rockland Immunochemicals
Anti-lambda light chain Goat Polyclonal Antibody
Catalog # ROCK609-1111
Supplier: Rockland Immunochemicals
Jump to:
Product Details & Documents
Suitable for immunoblotting (western or dot blot), ELISA, immunoelectron microscopy and immunochemistry as well as other antibody based enzymatic assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:1000 - 1:5000;
Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:1 mg
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- Gel shift:Yes
- ImmunoPrecipitation:Yes
- Form:Liquid (sterile filtered)
- ImmunoFluorescence:Yes
- Storage temperature:Cool
- ChIP:Yes
- Antigen name:lambda light chain
- Format:Antibody fraction
- Immunogen:Anti-Human lambda light chain antibody was produced by repeated immunization with Human lambda light chain molecule in goat.
- Storage buffer:0,125 M Sodium Borate, 0,075 M Sodium Chloride, 0,005 M EDTA, pH 8.0, 0,1% (w/v) Sodium Azide,
- Purification:purified by immunoaffinity chromatography
- Antibody type:Secondary
- Host:Goat
- Concentration:0,98 mg/ml by UV absorbance at 280 nm
- Cross adsorption:none
- Shipping temperature:Ice
- Cat. No.:ROCK609-1111
- Reactivity:Human
- Neutralisation:Yes
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- ELISA:Yes



