Anti-HUMAN TRANSFERRIN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK209-4234
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:transferrin
- Antigen symbol:TF
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:7018
- Antigen synonyms:PRO2086|PRO1557|TFQTL1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Polyethylene Glycol (PEG-8000)
- Storage temperature:Cool
- Concentration:10,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Transferrin (Human Serum)
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Size:20 mg
- Pk:20 mg
Specifications
About this item
Primary Rabbit Anti-HUMAN TRANSFERRIN Reacts with Human
Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: HUMAN TRANSFERRIN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human