Anti-Glutamate NMDA Receptor NR3A/B Subunit Mouse monoclonal antibody unconjugated
Catalog # QEDB60100
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Glutamate NMDA Receptor NR3A/B Subunit
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Glutamate NMDA Receptor NR3A/B Subunit
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat