Anti-ADAM22 Mouse Monoclonal Antibody
QEDB56528
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 22
- Antigen symbol:ADAM22
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Epitope:cytoplasmic domain
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: ADAM22 (ADAM metallopeptidase domain 22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Mouse
Isotype: IgG2b
Reactivity: