Order Entry
Export
ContactUsLinkComponent
Anti-DLG4 Mouse monoclonal antibody unconjugated
Anti-DLG4 Mouse monoclonal antibody unconjugated
  QEDB56457
Anti-DLG4 Mouse monoclonal antibody unconjugated
  QEDB56457

 

  • Antibody type:
    Primary
  • Antigen name:
    Discs, large homolog 4 (Drosophila)
  • Antigen symbol:
    DLG4
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: DLG4 (discs, large homolog 4 (Drosophila))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Mouse, Rat