Anti-Acetylated Lysine Mouse Monoclonal Antibody
Catalog # QEDB56455
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Acetyl-Lysine
- Antigen symbol:AcK
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Acetylated Lysine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: