Order Entry
Export
ContactUsLinkComponent
Anti-MAPK1/3 Rabbit Polyclonal Antibody
Anti-MAPK1/3 Rabbit Polyclonal Antibody
Catalog # QEDB56255
Supplier:  QED Bioscience
Anti-MAPK1/3 Rabbit Polyclonal Antibody
Catalog # QEDB56255
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 1/3
  • Antigen symbol:
    MAPK1/3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: