Anti-Insulin receptor (alpha subunit) Mouse Monoclonal Antibody
QEDB50101
- Antibody type:Primary
- Antigen name:Insulin receptor (alpha subunit)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Insulin receptor (alpha subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human