Anti-Interleukin-1 Receptor (IL-1R) Mouse Monoclonal Antibody
Catalog # QEDB40103
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-1 Receptor
- Antigen symbol:IL-1R
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Interleukin-1 Receptor (IL-1R)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human