Anti-Glucocorticoid Receptor Sheep Polyclonal Antibody
Catalog # QEDB36101
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Glucocorticoid Receptor
- Antigen symbol:GCR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Glucocorticoid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: