Anti-WWOX Rabbit Polyclonal Antibody
QEDB34016
- Antibody type:Primary
- Antigen name:WW domain containing oxidoreductase
- Antigen symbol:WWOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: WWOX (WW domain containing oxidoreductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human