Order Entry
Export
ContactUsLinkComponent
Anti-NKX3-1 Rabbit Polyclonal Antibody
Anti-NKX3-1 Rabbit Polyclonal Antibody
Catalog # QEDB34013
Supplier:  QED Bioscience
Anti-NKX3-1 Rabbit Polyclonal Antibody
Catalog # QEDB34013
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NK3 homeobox 1
  • Antigen symbol:
    NKX3-1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: NKX3-1 (NK3 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human